Gene Transcripts
- Gene Symbol
- Speer6-ps1
- Entrez Gene ID
- 73266
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 6, pseudogene 1
- Gene Aliases
- 1700039A02Rik, SPEER-6
- Chromosome Location
- Chr. 13: 3149475 - 3189678 on Build GRCm38
- UniGene ID
- Mm.335891
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001581.2 |
|
4, 5 |
458 |
|
GenBank mRNA |
AK006638.1 |
- |
4, 5 |
459 |
|
View Full Product Details