Gene Transcripts
- Gene Symbol
- Rian
- Entrez Gene ID
- 75745
- Gene Name
- RNA imprinted and accumulated in nucleus
- Gene Aliases
- 5530401N18Rik, C130089L09Rik, Meg8
- Chromosome Location
- Chr. 12: 109603945 - 109661716 on Build GRCm38
- UniGene ID
- Mm.293263
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028261.1 |
|
2, 3 |
255 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB195177.1 |
- |
2, 3 |
255 |
|
Chr. 12 - 109603945 - 109645375 [+] |
View Full Product Details