Gene Transcripts
- Gene Symbol
- CECR5-AS1
- Entrez Gene ID
- 100130717
- Gene Name
- CECR5 antisense RNA 1
- Gene Aliases
- CECR4, NCRNA00017
- Chromosome Location
- Chr. 22: 17159389 - 17165445 on Build GRCh38
- UniGene ID
- Hs.660066
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI962691.1 |
- |
1 |
44 |
|
Chr. 22 - 17159389 - 17165419 [+] |
GenBankmRNA |
CK903337.1 |
- |
1 |
60 |
|
Chr. 22 - 17159382 - 17165439 [+] |
View Full Product Details