Gene Transcripts

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
Chromosome Location
Chr. 3: 14124940 - 14143680 on Build GRCh38
UniGene ID
Hs.517817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017007176.1 XP_016862665.1 2 228
GenBank mRNA AK026869.1 - 1 1151

View Full Product Details