Gene Transcripts
- Gene Symbol
- RASEF
- Entrez Gene ID
- 158158
- Gene Name
- RAS and EF-hand domain containing
- Gene Aliases
- RAB45
- Chromosome Location
- Chr. 9: 82979585 - 83063177 on Build GRCh38
- UniGene ID
- Hs.129136
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057300.1 |
- |
1 |
1734 |
|
View Full Product Details