Gene Transcripts
- Gene Symbol
- ARHGEF1
- Entrez Gene ID
- 9138
- Gene Name
- Rho guanine nucleotide exchange factor 1
- Gene Aliases
- GEF1, LBCL2, LSC, P115-RHOGEF, SUB1.5
- Chromosome Location
- Chr. 19: 41883200 - 41907454 on Build GRCh38
- UniGene ID
- Hs.631550
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK026781.1 |
- |
1 |
693 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM041007.1 |
- |
1 |
196 |
|
Chr. 19 - 41889284 - 41890013 [+] |
GenBankmRNA |
BQ711445.1 |
- |
1 |
73 |
|
Chr. 19 - 41889406 - 41890281 [+] |
View Full Product Details