Gene Transcripts
- Gene Symbol
- BRD8
- Entrez Gene ID
- 10902
- Gene Name
- bromodomain containing 8
- Gene Aliases
- SMAP, SMAP2, p120
- Chromosome Location
- Chr. 5: 138139766 - 138178669 on Build GRCh38
- UniGene ID
- Hs.519337
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124191.1 |
- |
1 |
1921 |
|
View Full Product Details