Gene Transcripts
- Gene Symbol
- Msl3
- Entrez Gene ID
- 17692
- Gene Name
- male-specific lethal 3 homolog (Drosophila)
- Gene Aliases
- AU018931, Msl31, Msl3l1
- Chromosome Location
- Chr. X: 168651267 - 168674388 on Build GRCm38
- UniGene ID
- Mm.24248
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK136459.1 |
- |
1 |
2394 |
|
View Full Product Details