Gene Transcripts
- Gene Symbol
- Ccdc73
- Entrez Gene ID
- 211936
- Gene Name
- coiled-coil domain containing 73
- Gene Aliases
- 2210415I11Rik, 4931417A20, AV080665, BC095976
- Chromosome Location
- Chr. 2: 104886324 - 104999737 on Build GRCm38
- UniGene ID
- Mm.416538
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019067.1 |
- |
|
211 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY708825.1 |
- |
2 |
207 |
|
Chr. 2 - 105017147 - 105017906 [+] |
View Full Product Details