Gene Transcripts
- Gene Symbol
- Wwp1
- Entrez Gene ID
- 107568
- Gene Name
- WW domain containing E3 ubiquitin protein ligase 1
- Gene Aliases
- 8030445B08Rik, AIP5, SDRP1, Tiul1
- Chromosome Location
- Chr. 4: 19608296 - 19709004 on Build GRCm38
- UniGene ID
- Mm.474845
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK136811.1 |
- |
1 |
1895 |
|
View Full Product Details