Gene Transcripts
- Gene Symbol
- Als2cr11
- Entrez Gene ID
- 73463
- Gene Name
- amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11 (human)
- Gene Aliases
- 1700052H20Rik, 4930408G06Rik
- Chromosome Location
- Chr. 1: 59050506 - 59095015 on Build GRCm38
- UniGene ID
- Mm.252811
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015211.1 |
- |
|
265 |
|
AK019575.1 |
- |
|
375 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN834703.1 |
- |
|
148 |
|
Chr. - - [-] |
GenBankmRNA |
BY714657.1 |
- |
1 |
375 |
|
Chr. 1 - 58997326 - 59003539 [-] |
GenBankmRNA |
BY714766.1 |
- |
1 |
265 |
|
Chr. 1 - 58997117 - 59003429 [-] |
GenBankmRNA |
CN837599.1 |
- |
1 |
114 |
|
Chr. 1 - 58997142 - 59003278 [-] |
View Full Product Details