Gene Transcripts
- Gene Symbol
- Hotair
- Entrez Gene ID
- 100503872
- Gene Name
- HOX transcript antisense RNA (non-protein coding)
- Gene Aliases
- Gm16258, OTTMUSG00000032781
- Chromosome Location
- Chr. 15: 102944062 - 102947946 on Build GRCm38
- UniGene ID
- Mm.383980
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_047528.1 |
|
2 |
788 |
|
GenBank mRNA |
AK035706.1 |
- |
2 |
572 |
|
BB628325.1 |
- |
2 |
569 |
|
View Full Product Details