Gene Transcripts
- Gene Symbol
- Vps8
- Entrez Gene ID
- 209018
- Gene Name
- vacuolar protein sorting 8 homolog (S. cerevisiae)
- Gene Aliases
- AI315068, AU040738, mKIAA0804
- Chromosome Location
- Chr. 16: 21423118 - 21644681 on Build GRCm38
- UniGene ID
- Mm.235683
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285893.1 |
NP_001272822.1
|
42, 43 |
3998 |
|
NM_001285894.1 |
NP_001272823.1
|
42, 43 |
3861 |
|
GenBank mRNA |
AK029007.1 |
- |
26, 27 |
2215 |
|
AK031205.1 |
- |
19, 20 |
2040 |
|
AK122375.1 |
- |
42, 43 |
3810 |
|
AK141678.1 |
- |
42, 43 |
3921 |
|
AK219023.1 |
- |
2, 3 |
179 |
|
BC023914.1 |
- |
28, 29 |
2389 |
5323126 |
BC032214.1 |
- |
22, 23 |
1967 |
5323693 |
BC055323.1 |
- |
42, 43 |
3915 |
5706288 |
View Full Product Details