Gene Transcripts

Gene Symbol
Ubxn10
Entrez Gene ID
212190
Gene Name
UBX domain protein 10
Gene Aliases
5730509E04Rik, A830047D02, Ubxd3
Chromosome Location
Chr. 4: 138709837 - 138738438 on Build GRCm38
UniGene ID
Mm.292226
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285928.1 NP_001272857.1 4 996
NM_001285929.1 NP_001272858.1 3 885
NM_001285930.1 NP_001272859.1 3 749
NM_178671.5 NP_848786.1 2 618
GenBank mRNA AK045964.1 - 2 576
AK046949.1 - 4 997
AK077656.1 - 2 620
AK080662.1 - 2 584
AK133185.1 - 2 604
AK161245.1 - 2 605
AK161296.1 - 2 581
BC027099.1 - 3 749 5041000

View Full Product Details