Gene Transcripts

Gene Symbol
Slc10a5
Entrez Gene ID
241877
Gene Name
solute carrier family 10 (sodium/bile acid cotransporter family), member 5
Gene Aliases
Gm405, mP5
Chromosome Location
Chr. 3: 10331734 - 10335656 on Build GRCm38
UniGene ID
Mm.442146
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010834.2 NP_001010834.1 1 499
GenBank mRNA AK143900.1 - 1 500
AY825925.1 - 1 441
BC141377.1 - 1 476 9055990

View Full Product Details