Gene Transcripts

Gene Symbol
Lyg2
Entrez Gene ID
332427
Gene Name
lysozyme G-like 2
Gene Aliases
Gm776
Chromosome Location
Chr. 1: 37905583 - 37916493 on Build GRCm38
UniGene ID
Mm.332981
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033427.3 NP_001028599.1 5 748
GenBank mRNA AK132442.1 - 5 750
BC147562.1 - 5 685 9056552
BC147567.1 - 5 685 9056557

View Full Product Details