Gene Transcripts

Gene Symbol
Zhx2
Entrez Gene ID
387609
Gene Name
zinc fingers and homeoboxes 2
Gene Aliases
Afr-1, Afr1, Raf, mKIAA0854
Chromosome Location
Chr. 15: 57694667 - 57839832 on Build GRCm38
UniGene ID
Mm.36557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199449.2 NP_955520.1 3 674
GenBank mRNA AB099526.1 - 1 57
AK031782.1 - 3 571
AK157631.1 - 3 661
BC059178.1 - 3 428 6400319

View Full Product Details