Gene Transcripts

Gene Symbol
Rad21l
Entrez Gene ID
668929
Gene Name
RAD21-like (S. pombe)
Gene Aliases
Gm14160, MeiRAD21L, Rad21l1
Chromosome Location
Chr. 2: 151644366 - 151670622 on Build GRCm38
UniGene ID
Mm.473683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276400.1 NP_001263329.1 8 928
GenBank mRNA AB574185.1 - 8 928
AB602048.1 - 8 928
HQ603828.1 - 8 928

View Full Product Details