Gene Transcripts

Gene Symbol
Man2a1
Entrez Gene ID
25478
Gene Name
mannosidase, alpha, class 2A, member 1
Gene Aliases
Mana2, Manno
Chromosome Location
Chr. 9: 112293388 - 112451677 on Build Rnor_6.0
UniGene ID
Rn.163804
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012979.2 NP_037111.2 2, 3 1379
GenBank mRNA FQ225400.1 - 2, 3 559
FQ225693.1 - 2, 3 559
M24353.1 - 2, 3 296

View Full Product Details