Gene Transcripts

Gene Symbol
Prrx1
Entrez Gene ID
266813
Gene Name
paired related homeobox 1
Gene Aliases
Pmx1, Prx-1
Chromosome Location
Chr. 13: 81147038 - 81215560 on Build Rnor_6.0
UniGene ID
Rn.203584
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153821.1 NP_722543.1 2 684
XM_006250151.2 XP_006250213.1 2 1422
GenBank mRNA AY730764.1 - 2 408
S82911.1 - 2 684

View Full Product Details