Gene Transcripts

Gene Symbol
Lyzl6
Entrez Gene ID
287751
Gene Name
lysozyme-like 6
Gene Aliases
Lyzb, RGD1306968
Chromosome Location
Chr. 10: 91694547 - 91700915 on Build Rnor_6.0
UniGene ID
Rn.27809
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135833.1 NP_001129305.2 3 247
XM_006247503.2 XP_006247565.1 3 1760

View Full Product Details