Gene Transcripts

Gene Symbol
Mrps18c
Entrez Gene ID
289469
Gene Name
mitochondrial ribosomal protein S18C
Gene Aliases
Chromosome Location
Chr. 14: 10440691 - 10446909 on Build Rnor_6.0
UniGene ID
Rn.105741
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105996.1 NP_001099466.1 4 297
GenBank mRNA FQ224447.1 - 430

View Full Product Details