Gene Transcripts

Gene Symbol
Rgs7bp
Entrez Gene ID
294715
Gene Name
regulator of G-protein signaling 7 binding protein
Gene Aliases
R7bp, RGD1309360
Chromosome Location
Chr. 2: 35077300 - 35104743 on Build Rnor_6.0
UniGene ID
Rn.198108
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012347.1 NP_001012347.1 834
GenBank mRNA BC089977.1 - 834

View Full Product Details