Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100538.1 | NP_001094008.1 | 6 | 1193 | |
XM_006238798.1 | XP_006238860.1 | 6 | 1733 | ||
XM_006238799.1 | XP_006238861.1 | 6 | 1733 | ||
XM_006238800.1 | XP_006238862.1 | 6 | 1733 | ||
XM_006238801.1 | XP_006238863.1 | 6 | 1733 | ||
GenBank mRNA | BC088337.1 | - | 1 | 92 | |
BC166452.1 | - | 6 | 1198 | 8372968 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008769025.1 | XP_008767247.1 | 1 | 1167 |