Gene Transcripts
- Gene Symbol
- Otud6a
- Entrez Gene ID
- 302426
- Gene Name
- OTU deubiquitinase 6A
- Gene Aliases
- RGD1560855
- Chromosome Location
- Chr. X: 70256299 - 70257744 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_228584.3 |
XP_228584.1
|
1 |
679 |
|
View Full Product Details