Gene Transcripts

Gene Symbol
Sptbn1
Entrez Gene ID
305614
Gene Name
spectrin, beta, non-erythrocytic 1
Gene Aliases
Spnb1, Spnb2
Chromosome Location
Chr. 14: 114517839 - 114699645 on Build Rnor_6.0
UniGene ID
Rn.93208
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013130.1 NP_001013148.1 20 4339
XM_008770458.1 XP_008768680.1 20 5876
XM_008770460.1 XP_008768682.1 19 5733
XM_008770461.1 XP_008768683.1 20 4356
XM_008770462.1 XP_008768684.1 19 5733
XM_008770463.1 XP_008768685.1 20 5876
GenBank mRNA AY238344.1 - 20 4339

View Full Product Details