Gene Transcripts

Gene Symbol
Sephs2
Entrez Gene ID
308993
Gene Name
selenophosphate synthetase 2
Gene Aliases
Chromosome Location
Chr. 1: 198719158 - 198721476 on Build Rnor_6.0
UniGene ID
Rn.100471
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079889.2 NP_001073358.2 1 1269
GenBank mRNA BC081915.1 - 1 1164
BC097429.1 - 1 1269
BC128783.1 - 1 1126 7378558

View Full Product Details