Gene Transcripts

Gene Symbol
Mki67
Entrez Gene ID
291234
Gene Name
marker of proliferation Ki-67
Gene Aliases
Chromosome Location
Chr. 1: 207993895 - 208020454 on Build Rnor_6.0
UniGene ID
Rn.233802
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271366.1 NP_001258295.1 14 9145
XM_006230453.2 XP_006230515.1 13 9506
GenBank mRNA FQ227329.1 - 2 123
FQ230341.1 - 2 123

View Full Product Details