Gene Transcripts

Gene Symbol
Chpf
Entrez Gene ID
316533
Gene Name
chondroitin polymerizing factor
Gene Aliases
D1bwg1363e
Chromosome Location
Chr. 9: 82669199 - 82673898 on Build Rnor_6.0
UniGene ID
Rn.3794
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005906.1 NP_001005906.1 3 1242
XM_006245228.1 XP_006245290.1 3 871
GenBank mRNA BC083617.1 - 3 1242

View Full Product Details