Gene Transcripts

Gene Symbol
Nufip1
Entrez Gene ID
364430
Gene Name
nuclear fragile X mental retardation protein interacting protein 1
Gene Aliases
Chromosome Location
Chr. 15: 58171163 - 58201888 on Build Rnor_6.0
UniGene ID
Rn.154742
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007758.1 NP_001007759.1 7 827
GenBank mRNA BC082111.1 - 7 827

View Full Product Details