Gene Transcripts

Gene Symbol
Snrpc
Entrez Gene ID
361808
Gene Name
small nuclear ribonucleoprotein polypeptide C
Gene Aliases
Snrp1c, U1-C, U1C
Chromosome Location
Chr. 20: 7409401 - 7427581 on Build Rnor_6.0
UniGene ID
Rn.2786
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271040.1 NP_001257969.1 7 965
XM_006256220.2 XP_006256282.1 8 1024

Gene Symbol
LOC685273
Entrez Gene ID
685273
Gene Name
similar to U1 small nuclear ribonucleoprotein C (U1 snRNP protein C) (U1C protein) (U1-C)
Gene Aliases
Chromosome Location
Chr. 10: 14532653 - 14533400 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001063116.5 XP_001063116.2 1 642

View Full Product Details