Gene Transcripts

Gene Symbol
Mansc1
Entrez Gene ID
690606
Gene Name
MANSC domain containing 1
Gene Aliases
Chromosome Location
Chr. 4: 168363198 - 168382527 on Build Rnor_6.0
UniGene ID
Rn.230566
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109603.2 NP_001103073.1 3 608
XM_006237537.2 XP_006237599.1 3 735
XM_006237538.2 XP_006237600.1 3 740

View Full Product Details