Gene Transcripts

Gene Symbol
LOC102546874
Entrez Gene ID
102546874
Gene Name
prostate-specific antigen-like
Gene Aliases
Chromosome Location
Chr. 1: 99939243 - 99947079 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759477.1 XP_008757699.1 1 107

View Full Product Details