Gene Transcripts

Gene Symbol
CTXN1
Entrez Gene ID
404217
Gene Name
cortexin 1
Gene Aliases
CTXN
Chromosome Location
Chr. 19: 7924496 - 7926166 on Build GRCh38
UniGene ID
Hs.657978
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206833.3 NP_996664.1 2 960
GenBank mRNA AK098834.1 - 2 960
BC062702.1 - 2 942 4280819

View Full Product Details