Gene Transcripts

Gene Symbol
Krtap1-4
Entrez Gene ID
629873
Gene Name
keratin associated protein 1-4
Gene Aliases
OTTMUSG00000004966
Chromosome Location
Chr. 11: 99582549 - 99583677 on Build GRCm38
UniGene ID
Mm.423246
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039502.2 NP_001034591.1 1 1023
GenBank mRNA AK131904.1 - 1 1024

View Full Product Details