Gene Transcripts

Gene Symbol
Cmc2
Entrez Gene ID
66531
Gene Name
COX assembly mitochondrial protein 2
Gene Aliases
1110046L09Rik, 2310061C15Rik, DC13
Chromosome Location
Chr. 8: 116888685 - 116944209 on Build GRCm38
UniGene ID
Mm.4011
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026844.3 NP_081120.2 4 821
GenBank mRNA AK009995.1 - 4 852
AK010073.1 - 4 852
AK013475.1 - 4 818

View Full Product Details