Gene Transcripts
- Gene Symbol
- CNPY3
- Entrez Gene ID
- 10695
- Gene Name
- canopy FGF signaling regulator 3
- Gene Aliases
- CAG4A, ERDA5, PRAT4A, TNRC5
- Chromosome Location
- Chr. 6: 42928002 - 42939287 on Build GRCh38
- UniGene ID
- Hs.414099
- Species
- Homo sapiens
- Gene Symbol
- CNPY3-GNMT
- Entrez Gene ID
- 107080644
- Gene Name
- CNPY3-GNMT readthrough
- Gene Aliases
- Chromosome Location
- Chr. 6: 42929130 - 42963880 on Build GRCh38
- UniGene ID
- Hs.144914
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_134890.1 |
|
2 |
609 |
|
GenBank mRNA |
FY213816.1 |
- |
2 |
141 |
|
View Full Product Details