Gene Transcripts

Gene Symbol
Ndufa10
Entrez Gene ID
678759
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10
Gene Aliases
ndufa10l
Chromosome Location
Chr. 9: 99617051 - 99651825 on Build Rnor_6.0
UniGene ID
Rn.169414
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199495.2 NP_955789.2 2 259
XM_008767385.1 XP_008765607.1 2 271
GenBank mRNA BC093375.1 - 2 218
CB610197.1 - 2 259
FQ209525.1 - 2 261

Gene Symbol
Ndufa10l1
Entrez Gene ID
316632
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10-like 1
Gene Aliases
Ndufa10
Chromosome Location
Chr. 6: 62796951 - 62798384 on Build Rnor_6.0
UniGene ID
Rn.102304
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182671.2 NP_872612.1 1 221
GenBank mRNA AY260163.1 - 1 221

View Full Product Details