Gene Transcripts

Gene Symbol
LCE1C
Entrez Gene ID
353133
Gene Name
late cornified envelope 1C
Gene Aliases
LEP3
Chromosome Location
Chr. 1: 152804835 - 152806631 on Build GRCh38
UniGene ID
Hs.516429
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276331.1 NP_001263260.1 3 325
NM_178351.3 NP_848128.1 2 415

View Full Product Details