Gene Transcripts

Gene Symbol
MRPS18C
Entrez Gene ID
51023
Gene Name
mitochondrial ribosomal protein S18C
Gene Aliases
CGI-134, MRP-S18-1, MRP-S18-c, MRPS18-1, S18mt-c, mrps18-c
Chromosome Location
Chr. 4: 83455932 - 83461723 on Build GRCh38
UniGene ID
Hs.334772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016067.3 NP_057151.1 3, 4 383
GenBank mRNA AF151892.1 - 3, 4 296
AK026620.1 - 3, 4 350
BC005186.1 - 3, 4 274 3839381

View Full Product Details