Gene Transcripts

Gene Symbol
LYZL6
Entrez Gene ID
57151
Gene Name
lysozyme like 6
Gene Aliases
HEL-S-6a, LYC1, LYZB, PRO1485, TKAL754, UNQ754
Chromosome Location
Chr. 17: 35934518 - 35943710 on Build GRCh38
UniGene ID
Hs.97477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199951.2 NP_001186880.1 3, 4 529
NM_020426.3 NP_065159.1 3, 4 643
GenBank mRNA AY359018.1 - 2, 3 447
AY742214.1 - 2, 3 419
BC054481.1 - 3, 4 489 6620264
BG195727.1 - 2, 3 497
GQ472214.1 - 3, 4 489

View Full Product Details