Gene Transcripts

Gene Symbol
Srxn1
Entrez Gene ID
296271
Gene Name
sulfiredoxin 1
Gene Aliases
Ab2-390, Npn3
Chromosome Location
Chr. 3: 147603969 - 147632824 on Build Rnor_6.0
UniGene ID
Rn.2835
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047858.3 NP_001041323.3 2 520
XM_008762308.1 XP_008760530.1 3 315
GenBank mRNA AY325205.1 - 3 315
BC167743.1 - 2 273

View Full Product Details