Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081003.2 | NP_001074472.1 | 2 | 406 | |
NM_001081004.2 | NP_001074473.1 | 2 | 274 | ||
NM_001287237.1 | NP_001274166.1 | 2 | 541 | ||
NM_014066.3 | NP_054785.2 | 2 | 428 | ||
GenBank mRNA | AF113540.1 | - | 2 | 559 | |
AF290195.1 | - | 2 | 436 | ||
AK001477.1 | - | 2 | 254 | ||
AK023070.1 | - | 2 | 375 | ||
BC002672.2 | - | 2 | 258 | 3606279 | |
BC003055.1 | - | 2 | 346 | 3544106 | |
BC065729.1 | - | 2 | 396 | 6452461 | |
BG705627.1 | - | 2 | 275 | ||
BI753202.1 | - | 2 | 323 | ||
BM013651.1 | - | 2 | 477 | ||
BM924244.1 | - | 2 | 388 | ||
BQ680339.1 | - | 270 | |||
BU508224.1 | - | 2 | 347 | ||
BU600590.1 | - | 392 |