Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077394.1 | NP_001070862.1 | 2 | 270 | |
NM_001077395.1 | NP_001070863.1 | 2 | 252 | ||
NM_015958.2 | NP_057042.2 | 2 | 252 | ||
XM_005270938.1 | XP_005270995.1 | 2 | 252 | ||
XM_005270941.3 | XP_005270998.1 | 2 | 187 | ||
XM_011541569.1 | XP_011539871.1 | 2 | 252 | ||
XM_017001460.1 | XP_016856949.1 | 2 | 272 | ||
XM_017001462.1 | XP_016856951.1 | 2 | 209 | ||
GenBank mRNA | AF132964.1 | - | 2 | 221 | |
AF157319.1 | - | 2 | 220 | ||
AF161492.1 | - | 2 | 261 | ||
AF248965.1 | - | 2 | 221 | ||
AK125485.1 | - | 2 | 226 | ||
AK225469.1 | - | 2 | 230 | ||
AK225707.1 | - | 2 | 230 | ||
AK225951.1 | - | 2 | 230 | ||
AK289351.1 | - | 2 | 241 | ||
AK298181.1 | - | 2 | 230 | ||
BC053857.1 | - | 2 | 218 | 5503038 | |
BI907367.1 | - | 2 | 225 | ||
CK002046.1 | - | 2 | 273 | ||
DA133565.1 | - | 2 | 252 |