Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160167.1 | NP_001153639.1 | 3 | 617 | |
NM_001160169.1 | NP_001153641.1 | 2 | 353 | ||
NM_024841.4 | NP_079117.3 | 4 | 603 | ||
GenBank mRNA | AK055042.1 | - | 3 | 504 | |
BX640959.1 | - | 4 | 584 | ||
DA344861.1 | - | 3 | 504 | ||
DR001466.1 | - | 2 | 286 |