Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001202855.2 | NP_001189784.1 | 8 | 801 | |
NM_017460.5 | NP_059488.2 | 8 | 804 | ||
XM_017011780.1 | XP_016867269.1 | 8 | 802 | ||
GenBank mRNA | AF182273.1 | - | 8 | 706 | |
AK298451.1 | - | 7 | 675 | ||
AK298462.1 | - | 6 | 551 | ||
AK312967.1 | - | 8 | 797 | ||
AY606313.2 | - | 8 | 699 | ||
BC069418.1 | - | 8 | 747 | 7262313 | |
BC101631.1 | - | 8 | 793 | 8069137 | |
D00003.1 | - | 8 | 757 | ||
DQ924960.1 | - | 8 | 697 | ||
J04449.1 | - | 8 | 762 | ||
M13785.1 | - | 8 | 757 | ||
M14096.1 | - | 8 | 765 | ||
M18907.1 | - | 8 | 787 | ||
X12387.1 | - | 8 | 766 |