Gene Transcripts

Gene Symbol
OR2M3
Entrez Gene ID
127062
Gene Name
olfactory receptor family 2 subfamily M member 3
Gene Aliases
OR1-54, OR2M3P, OR2M6, OST003
Chromosome Location
Chr. 1: 248203068 - 248204006 on Build GRCh38
UniGene ID
Hs.553581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004689.1 NP_001004689.1 1 406
GenBank mRNA BC136970.1 - 1 444
BC136971.1 - 1 444 9020963

View Full Product Details