Gene Transcripts

Gene Symbol
CWF19L2
Entrez Gene ID
143884
Gene Name
CWF19-like 2, cell cycle control (S. pombe)
Gene Aliases
Chromosome Location
Chr. 11: 107326345 - 107457846 on Build GRCh38
UniGene ID
Hs.212140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152434.2 NP_689647.2 16 2421
XM_017017239.1 XP_016872728.1 15 1739
GenBank mRNA AK056905.1 - 14 2073
AK126881.1 - 16 2412
AK293001.1 - 16 2356
BC028043.2 - 3 270
BC056241.1 - 4 516
BC110439.1 - 14 2111 40001346
BC110440.1 - 13 2033 40001348
BC110441.1 - 13 1976 40001349
BC110442.1 - 13 2034 40001350
BC118669.1 - 13 2037 40012545
BX647832.1 - 16 2395
BX648704.1 - 16 2400
BX649022.1 - 16 2391

View Full Product Details