Gene Transcripts
- Gene Symbol
- OTUD7A
- Entrez Gene ID
- 161725
- Gene Name
- OTU deubiquitinase 7A
- Gene Aliases
- C15orf16, C16ORF15, CEZANNE2, OTUD7
- Chromosome Location
- Chr. 15: 31483126 - 31655339 on Build GRCh38
- UniGene ID
- Hs.355236
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_130901.1 |
NP_570971.1
|
11 |
3000 |
|
GenBank mRNA |
AJ430383.1 |
- |
11 |
3000 |
|
View Full Product Details