Gene Transcripts

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
Chromosome Location
Chr. 3: 58716415 - 59050078 on Build GRCh38
UniGene ID
Hs.368434
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198463.2 NP_940865.1 9, 10 1041
XM_006713013.3 XP_006713076.1 9, 10 1421
XM_011533451.2 XP_011531753.1 8, 9 1256
XM_011533453.2 XP_011531755.1 9, 10 1421
XM_011533455.2 XP_011531757.1 9, 10 1421
XM_011533456.1 XP_011531758.1 9, 10 1130
XM_011533457.1 XP_011531759.1 7, 8 910
XM_011533458.2 XP_011531760.1 6, 7 793
XM_011533459.2 XP_011531761.1 9, 10 1421
XM_011533460.2 XP_011531762.1 4, 5 560
XM_011533461.1 XP_011531763.1 6, 7 753
XM_011533463.2 XP_011531765.1 3, 4 384
XM_017005848.1 XP_016861337.1 5, 6 666
XM_017005849.1 XP_016861338.1 8, 9 1264
XM_017005850.1 XP_016861339.1 8, 9 942
GenBank mRNA AK124111.1 - 9, 10 1043
AK124920.1 - 1, 2 133
BC050317.1 - 9, 10 1429
BC132815.1 - 7, 8 855 40146837
BC151142.1 - 7, 8 855 9020498

View Full Product Details